Learn More
Lonza™ BioWhittaker™ Phosphate Buffered Solution (1X)
Serves as a wash solution in a wide variety of laboratory applications. BioWhittaker™ Phosphate Buffered Solution (1X) Phosphate Buffered Saline is manufactured in accordance with cGMP regulations.
Brand: Lonza™ BE17-516Q
This item is not returnable.
View return policy
Description
- Wash solution
- Manufactured in accordance with cGMP regulations
- Cell culture tested to assure compatibility
Specifications
No Calcium, No Magnesium | |
Phosphate Buffered Saline (PBS) | |
7.3 to 7.5 |
15° to 30°C | |
1 L | |
Sterile |