Learn More
BioWhittaker™ Water for Cell Culture, Lonza™
Click to view available options
Quantity:
500 mL
1 L
Description
- Method for the production of WFI includes:
- Ultra filtration
- Reverse osmosis
- Mixed bed deionizers
- Hot recirculating loop and cooled at point of use
- System is validated and monitored for bioburden, endotoxin, total organic carbons and conductivity
Specifications
Specifications
Sterility | Sterile |
Quantity | 1 L |
Chemical Name or Material | Water |