Learn More
Bis(cyclopentadienyl)manganese, sublimed, Thermo Scientific Chemicals
788.93 EUR
Chemical Identifiers
CAS | 73138-26-8 |
---|---|
Molecular Formula | C10H10Mn |
Molecular Weight (g/mol) | 185.13 |
MDL Number | MFCD00045468 |
InChI Key | LCGVCXIFXLGLHG-UHFFFAOYSA-N |
Synonym | bis cyclopentadienyl manganese, acmc-20aln3, dicyclopentadienylmanganese, manganese,di-2,4-cyclopentadien-1-yl, bis cyclopenta-1,3-dien-1-yl manganese |
PubChem CID | 21960473 |
IUPAC Name | cyclopenta-1,3-diene;manganese(2+) |
SMILES | [Mn](c1cccc1)c1cccc1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
73138-26-8 | |
185.13 | |
LCGVCXIFXLGLHG-UHFFFAOYSA-N | |
21960473 | |
[Mn](c1cccc1)c1cccc1 |
C10H10Mn | |
MFCD00045468 | |
bis cyclopentadienyl manganese, acmc-20aln3, dicyclopentadienylmanganese, manganese,di-2,4-cyclopentadien-1-yl, bis cyclopenta-1,3-dien-1-yl manganese | |
cyclopenta-1,3-diene;manganese(2+) |
Specifications
73138-26-8 | |
245°C | |
Ampule | |
MFCD00045468 | |
Air and moisture sensitive | |
LCGVCXIFXLGLHG-UHFFFAOYSA-N | |
cyclopenta-1,3-diene;manganese(2+) | |
21960473 | |
Powder |
175°C | |
52°C (126°F) | |
C10H10Mn | |
UN3181 | |
bis cyclopentadienyl manganese, acmc-20aln3, dicyclopentadienylmanganese, manganese,di-2,4-cyclopentadien-1-yl, bis cyclopenta-1,3-dien-1-yl manganese | |
[Mn](c1cccc1)c1cccc1 | |
185.13 | |
185.13 | |
Bis(cyclopentadienyl)manganese, Sublimed |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 215-038-5
missing translation for 'tsca' : No
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.