missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(trimethylsilyl)acetylene, 99%, Thermo Scientific Chemicals
67.50 EUR - 796.00 EUR
Chemical Identifiers
CAS | 14630-40-1 |
---|---|
Molecular Formula | C8H18Si2 |
Molecular Weight (g/mol) | 170.41 |
MDL Number | MFCD00008276 |
InChI Key | ZDWYFWIBTZJGOR-UHFFFAOYSA-N |
Synonym | bis trimethylsilyl acetylene, silane, 1,2-ethynediylbis trimethyl, 1,2-bis trimethylsilyl ethyne, btmsa, 1,2-bis-trimethylsilanyl-ethyne, ethyne-1,2-diylbis trimethylsilane, 2,5-disilahex-3-yne, 2,2,5,5-tetramethyl, trimethyl 2-trimethylsilyl ethynyl silane, silane, 1,1'-1,2-ethynediyl bis 1,1,1-trimethyl |
PubChem CID | 84564 |
IUPAC Name | trimethyl(2-trimethylsilylethynyl)silane |
SMILES | C[Si](C)(C)C#C[Si](C)(C)C |
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
10206050
|
Thermo Scientific Acros
182010100 |
10 g | Glass bottle |
67.50 EUR
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10327270
|
Thermo Scientific Acros
182010500 |
50 g | Glass bottle |
246.00 EUR
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15119223
|
Thermo Scientific Acros
182012500 |
250 g | Glass bottle | ||||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
14630-40-1 | |
170.41 | |
ZDWYFWIBTZJGOR-UHFFFAOYSA-N | |
84564 | |
C[Si](C)(C)C#C[Si](C)(C)C |
C8H18Si2 | |
MFCD00008276 | |
bis trimethylsilyl acetylene, silane, 1,2-ethynediylbis trimethyl, 1,2-bis trimethylsilyl ethyne, btmsa, 1,2-bis-trimethylsilanyl-ethyne, ethyne-1,2-diylbis trimethylsilane, 2,5-disilahex-3-yne, 2,2,5,5-tetramethyl, trimethyl 2-trimethylsilyl ethynyl silane, silane, 1,1'-1,2-ethynediyl bis 1,1,1-trimethyl | |
trimethyl(2-trimethylsilylethynyl)silane |
Spezifikation
14630-40-1 | |
Colorless | |
136.0°C to 137.0°C | |
Authentic | |
C8H18Si2 | |
1.4260 to 1.4280 | |
05,44; 13,97 | |
bis trimethylsilyl acetylene, silane, 1,2-ethynediylbis trimethyl, 1,2-bis trimethylsilyl ethyne, btmsa, 1,2-bis-trimethylsilanyl-ethyne, ethyne-1,2-diylbis trimethylsilane, 2,5-disilahex-3-yne, 2,2,5,5-tetramethyl, trimethyl 2-trimethylsilyl ethynyl silane, silane, 1,1'-1,2-ethynediyl bis 1,1,1-trimethyl | |
ZDWYFWIBTZJGOR-UHFFFAOYSA-N | |
trimethyl(2-trimethylsilylethynyl)silane | |
84564 | |
99% | |
Bis(trimethylsilyl)acetylene, 99% |
21.0°C to 24.0°C | |
0.7500g/mL | |
2°C | |
98.5% min. (GC) | |
(CH3)3SiC=CSi(CH3)3 | |
MFCD00008276 | |
0.75 | |
Solubility in water: may decompose | |
C[Si](C)(C)C#C[Si](C)(C)C | |
170.41 | |
170.41 | |
Liquid After Melting |
Sicherheit und Handhabung
GHS H Statement
Flammable solid.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 238-671-9
missing translation for 'tsca' : TSCA
Produktidentifikator
Signalwort
Gefahrenkategorie
Gefahrenhinweis
Sicherheitshinweis
- Bis(trimethylsilyl)acetylene
Signalwort
- Danger
Gefahrenkategorie
- Flammable solid Category 1
Gefahrenhinweis
- H228-Flammable solid.
Sicherheitshinweis
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only