Learn More
Bis(triphenylphosphine)iminium trifluoroacetate, 98%, Thermo Scientific™
80.97 EUR - 362.00 EUR
Chemical Identifiers
CAS | 116405-43-7 |
---|---|
Molecular Formula | C38H30F3NO2P2 |
Molecular Weight (g/mol) | 651.61 |
MDL Number | MFCD15146355 |
InChI Key | NDLHFNFVQBXEPE-UHFFFAOYSA-M |
Synonym | 1,1,1-triphenyl-n-triphenylphosphoranylidene phosphoraniminium 2,2,2-trifluoroacetate, ppn tfa, ksc924o3n, bis triphenylphosphine iminiumtrifluoroacetate, bis-triphenylphosphoranylidene ammonium tfa, bis triphenylphosphine iminium trifluoroacetic acid salt, bis-triphenylphosphoranylidene ammonium tfa ppn tfa, bis triphenyl-??-phosphanylidene azanium trifluoroacetate, 1,1,1-triphenyl-n-triphenylphosphoranylidene-phosphoraniminium 2,2,2-trifluoroacetate, triphenyl-n-triphenyl-lambda∼5∼-phosphanylidene-lambda∼5∼-phosphaniminium trifluoroacetate |
PubChem CID | 19800386 |
IUPAC Name | 2,2,2-trifluoroacetate;triphenyl-[(triphenyl-$l^{5}-phosphanylidene)amino]phosphanium |
SMILES | [O-]C(=O)C(F)(F)F.C1=CC=C(C=C1)P(=[N+]=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsBis(triphenylphosphine)iminium trifluoroacetate is used as pharmaceutical intermediates and chemical intermediate.
Solubility
Soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Chemical Identifiers
116405-43-7 | |
651.61 | |
NDLHFNFVQBXEPE-UHFFFAOYSA-M | |
19800386 | |
[O-]C(=O)C(F)(F)F.C1=CC=C(C=C1)P(=[N+]=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C38H30F3NO2P2 | |
MFCD15146355 | |
1,1,1-triphenyl-n-triphenylphosphoranylidene phosphoraniminium 2,2,2-trifluoroacetate, ppn tfa, ksc924o3n, bis triphenylphosphine iminiumtrifluoroacetate, bis-triphenylphosphoranylidene ammonium tfa, bis triphenylphosphine iminium trifluoroacetic acid salt, bis-triphenylphosphoranylidene ammonium tfa ppn tfa, bis triphenyl-??-phosphanylidene azanium trifluoroacetate, 1,1,1-triphenyl-n-triphenylphosphoranylidene-phosphoraniminium 2,2,2-trifluoroacetate, triphenyl-n-triphenyl-lambda∼5∼-phosphanylidene-lambda∼5∼-phosphaniminium trifluoroacetate | |
2,2,2-trifluoroacetate;triphenyl-[(triphenyl-$l^{5}-phosphanylidene)amino]phosphanium |
Specifications
116405-43-7 | |
C38H30F3NO2P2 | |
1,1,1-triphenyl-n-triphenylphosphoranylidene phosphoraniminium 2,2,2-trifluoroacetate, ppn tfa, ksc924o3n, bis triphenylphosphine iminiumtrifluoroacetate, bis-triphenylphosphoranylidene ammonium tfa, bis triphenylphosphine iminium trifluoroacetic acid salt, bis-triphenylphosphoranylidene ammonium tfa ppn tfa, bis triphenyl-??-phosphanylidene azanium trifluoroacetate, 1,1,1-triphenyl-n-triphenylphosphoranylidene-phosphoraniminium 2,2,2-trifluoroacetate, triphenyl-n-triphenyl-lambda∼5∼-phosphanylidene-lambda∼5∼-phosphaniminium trifluoroacetate | |
NDLHFNFVQBXEPE-UHFFFAOYSA-M | |
2,2,2-trifluoroacetate;triphenyl-[(triphenyl-$l^{5}-phosphanylidene)amino]phosphanium | |
19800386 | |
98% | |
Bis(triphenylphosphine)iminium trifluoroacetate |
White | |
MFCD15146355 | |
Soluble in water. | |
[O-]C(=O)C(F)(F)F.C1=CC=C(C=C1)P(=[N+]=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
651.61 | |
782.65 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.