Learn More
Scientific Industries SI™ Blank Foam Inserts
Allows users to accommodate odd-shaped vessels. Scientific Industries SI™ Blank Foam Inserts are made of high-density foam that fits into a Vortex-Genie 2 Recessed Platform.
Brand: Scientific Industries SI™ 503-0277-00
Additional Details : Weight : 0.03420kg
Description
- Constructed of high-density foam
- Customizable for odd-shaped vessels.
- Fits into the Recessed Platform
- For use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.
- One insert is included in the Large Sample Set
Specifications
High-density foam | |
Vortex-Genie 2 Recessed Platform |