Learn More
Bollé Safety Over-the-Glass Safety Glasses
Enjoy guaranteed maximum safety for all, including wearers of prescription glasses, with Bollé Safety™ Over-the-Glass Safety Glasses. Made from polycarbonate in a highly wrap-around and attractive style.
10.10 EUR - 17.55 EUR
Specifications
Product Type | Over-the-Glass Safety Glasses |
---|---|
Lens Material | Polycarbonate |
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
13128766
|
Bollé Safety
OVLITLPSI |
33 g |
11.13 EUR
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
11745677
|
Bollé Safety
COVARSI |
68 g |
10.10 EUR
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
11755677
|
Bollé Safety
COVERSI |
67 g |
17.55 EUR
Each |
Estimated Shipment: 25-06-2025
Log in to see stock. |
|||||
Description
- Highly wrap-around frame
- Side, upper and lower protection
-
Specifications
Over-the-Glass Safety Glasses | |
Polycarbonate |