Learn More
BRAND™ Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Glass Stopper, with Certificate
Shop All Brand Products
Click to view available options
Capacity (Metric):
25 mL
100 mL
5 L
Unit Size:
Each
Description
- Class A Volumetric Flask
- Constructed of Borosilicate Glass
- Complies with DIN EN ISO 1042 standards
- Calibrated to contain (TC, In)
- Narrow neck and wide neck options available in a variety of sizes
- DE-M marking provides quick and accurate volume check
- Glass stopper included
- Delivered with individual conformity certificate
Specifications
Specifications
| Calibration | Calibrated To Contain |
| Certifications/Compliance | DIN EN ISO 1042 |
| Closure Material | Glass |
| Color | Clear |
| Description | DE-M Marking |
| Capacity (Metric) | 25 mL |
| Material | Borosilicate Glass |
| Limit of Error | ±0.06 mL |
| Neck Style | Wide |
| No. per Case | 2 Pack |
| Show More |