Learn More
Thermo Scientific Blood Bag Adapter
Blood Bag Adapter for 450 to 500 mL quad, triple blood bag XL (set of 6) for double blood bucket 75007617.
Brand:  Thermo Scientific 76007657
686.47 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Product Code. 11861471
  • 722.60 EUR / Pack of 6
Description
  • Adapter for 450 to 500 mL quad, triple blood bag ''XL'' (set of 6) for double blood bucket 75007617.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Blood Bag Adapter
450 to 500 mL
Quad, Triple Blood Bag ''XL'' (set of 6), Double Blood Bucket
Plastic