Learn More
Fisherbrand™ Color-coded Micro PTFE Stir Bar
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
2 mm
5 mm
7 mm
8 mm
10 mm
15 mm
Diameter (Metric):
1.5 mm
2 mm
3 mm
Color:
Blue
Red
Yellow
Unit Size:
Pack of 10
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | Blue |
| Description | PTFE encapsulated Alnico V magnet core |
| For Use With (Equipment) | Containers & Vessels / Stirrer Hot Plates |
| Diameter (Metric) | 2 mm |
| Length (Metric) | 2 mm |
| Quantity | 10/Pk. |
| Shape | Micro |