Learn More
SI Analytics™ BlueLine Plug Cable Combination
SI Analytics™ BlueLine Plug Cable Combination is a coaxial electrode plug that is designed to be used with SI Analytics™ BlueLine series.
Brand: SI Analytics™ 285122678
This item is not returnable.
View return policy
Description
- For use with BlueLine series
- Coaxial cable
- Available in 1m or 3m lengths
Specifications
Coaxial Electrode Plug | |
LB 3 A | |
3 m |
BlueLine | |
3m Coaxial Cable, DIN Instrument Plug |