Learn More
Brady™ BMP51/53 Labelmaker Labels, Rectangle
Popular wire and cable labels.
Brand:  Brady™ 143271
 View more versions of this product
Product Code. 11922792
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Conformable material designed to adhere to highly curved surfaces, wires and cables
  • Features a white printable area with clear 'tail' that wraps around marker
  • Clear tails serves as over-laminate to protect the text, providing superior resistance to abrasion, dirt, oils, solvents, water and excessive handling
Specifications
Specifications
Black on White/Transparent
38.10 mm
0.102 mm
Self-laminating BMP51/53 Labelmaker Labels
Rectangle
UL Recognized to UL969 Labelling and Marking Standard when printed with the Brady Series R4300 and R6200 ribbons. See UL files MH17154 for specific details. Brady customers should specify at purchase if they require UL Recognized B-427 labels. Brady sells both UL Recognized B-427 labels and non-approved labels depending on customer requirements.
Vinyl
101.60 mm
Acrylic
Matte
-70°C to 70°C
70/Pk.