Learn More
Brady™ BMP71 BMP61 M611 Vinyl General Identification Labels
Labels for the BMP61 and M611 Thermal Transfer Portable Printers.
Brand: Brady™ 011181
This item is not returnable.
View return policy
Description
- Can also be printed on the BMP™71 Label Printer provided the roll adapter (M71-ADAPT) is used
Specifications
Permanent Acrylic | |
Gloss | |
Thermal Transfer | |
15.24 m x 48.26 mm |
Blue | |
Polyester (B-439) | |
BMP61, BMP71 (with Media Adapter), M611 |