Learn More
Bohlender™ BOLA™ Flat Flange Lids
Flat Flange Lids suitable for reaction vessels with SCHOTT flat flange.
Brand:  Bohlender™ B210-14
 View more versions of this product
Product Code. 17156964
  • 793.00 EUR / Each
Description
  • Suitable for reaction vessels with SCHOTT flat flange
  • Center neck socket NS 29/32
  • Lateral necks with ground joint sockets or GL-thread
  • Bottom side with centring collar for non-slipping alignment on the vessel
  • All lateral connectors are made to insert tubing, tubes and probes either aside the center or vertical (parallel to the stirrer shaft) in order to avoid collisions
Specifications
Specifications
PTFE
Reaction Vessels with SCHOTT Flat Flange
DN 100
Lateral Necks: 3 x NS 29/32 Angled Aside the Shaft