missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ Screw In Stirrer Bearings

Screw In Stirrer Bearings includes screw cap and PTFE gasket.

Brand:  Bohlender™ B155-06

Product Code. 17166954

  • 155.00 EUR / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For screw in into the center neck of BOLA™ Reactor Lids for centric guidance of stirrer shafts
  • A special gasket made of PTFE and an FPM o-ring which is compressed by a GL screw cap made of PPS provide a good sealing of the stirrer shaft
  • With hexagonal gripping surface in standard wrench size
  • Universal chemical resistance
  • Medium is only exposed to PTFE
Specifications

Specifications

Stirrer Shafts with 6 mm Diameter
Wrench Size 25 mm
PTFE
GL 25, Screw-in Thread NPT 1/2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.