Learn More
Boric Acid 1%, Chem Lab™
Product Code. 11949162
Click to view available options
Quantity:
5 L
Unit Size:
5 L
Product Code. 11949162
Brand: Chem-Lab CL02.0202.5000
Specifications
pH ±4
Color Colorless
Density 1g/mL
Odor Odorless
Quantity 5 L
Physical Form Liquid
Solubility Information Water
compliance-icons
Product Identifier
  • Boric acid 1% solution
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Boric acid