Learn More
Pyrex™ Borosilicate Glass Automatic Burette with Intermediate Stopcock
Product Code. 12822104
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Graduations:
0.02 mL
0.05 mL
0.1 mL
Unit Size:
Each
Product Code. 12822104
Brand: Pyrex 3496/02AS
Only null left
Add to basket
Add to basket
  • As 3396 range with intermediate stopcock
  • Intermediate stopcock facilitates easy drainage of liquid back into reservoir
  • 1.5mm bore PTFE key stopcock
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 10 mL
Graduations 0.02 mL