Learn More
Fisherbrand™ Test Tubes, Borosilicate Glass 3.3, With Beaded Rim, heavy wall
Product Code. p-10209543 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
14 mL
47 mL
Unit Size:
Pack of 100
Pack of 50
Product Code. 15852295
Only null left
Add to basket
Add to basket
Manufactured from borosilicate glass with a heavy wall design for strong resistance to heat and chemicals. Fisherbrand™ Borosilicate Glass Heavy Wall Rimmed Test Tubes are ideal for chemical and biological work.
  • Features: rimmed heavy walls, superior mechanical strength
  • Grade: 3.3 borosilicate glass test tubes
  • Complies to: ISO 4142
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 4142
Capacity (Metric) 47 mL
Length (Metric) 150 mm
Material Borosilicate Glass
Quantity 50 Pack
Rimmed Yes
Diameter (Metric) 24 mm
Wall Thickness 1.8 mm