Learn More
Kimble Chase™ Glass Shell Vial with Closure
Kimble Chase™ Glass Shell Vial with Closure features an open-bottom plug style plastic closure.
Brand: Kimble Chase™ 60965D-7
This item is not returnable.
View return policy
Description
- TITESEAL™ vials
- Plain tops
- Open-bottom plug style plastic closures, unattached
- Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Specifications
432 | |
144 | |
Open Bottom Plug |