Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug, Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug, Scale are manufactured from durable DURAN™ glass.
75.00 EUR - 130.00 EUR
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11886204
|
Lenz Laborglasinstrumente™
04012073 |
2000 mL |
130.00 EUR
Each |
Estimated Shipment: 17-04-2025
Log in to see stock. |
|||||
11812652
|
Lenz Laborglasinstrumente™
04012070 |
1000 mL |
126.00 EUR
Pack of 2 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||||
11882642
|
Lenz Laborglasinstrumente™
04012037 |
100 mL |
75.00 EUR
Pack of 2 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||||
11802652
|
Lenz Laborglasinstrumente™
04012058 |
500 mL |
103.00 EUR
Pack of 2 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||||
11892642
|
Lenz Laborglasinstrumente™
|
250 mL |
84.00 EUR
Pack of 2 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||||
11872642
|
Lenz Laborglasinstrumente™
04012028 |
50 mL |
78.00 EUR
Pack of 2 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||||
Description
- With NS socket and PE stopper
- Scale of approx. volume/mL
- With screwthread retaining device
Specifications
Borosilicate Glass | |
Polyethylene |