Learn More
Emma Safety Footwear Boston Safety Shoes
The TPU GripForce™ Easy Twist outsole offers maximum grip, is durable and highly recyclable. The integrated lacing prevents pressure points from hooks and eyes and the Hydro-Tec™ lining provides optimal breathability.
93.45 EUR - 133.00 EUR
Specifications
Certifications/Compliance | EN ISO 20345:2011 |
---|---|
Closure Type | Laces |
Color | Black, Gray |
Design | Boston |
Liner Material | Hydro-Tec™ Antibacterial |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15978381
|
Emma Safety Footwear
MM40064739 |
39 |
109.18 EUR
1 pair |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
15988381
|
Emma Safety Footwear
MM40064740 |
40 |
125.08 EUR
1 pair |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
15998381
|
Emma Safety Footwear
MM40064741 |
41 |
133.00 EUR
1 pair |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
15908391
|
Emma Safety Footwear
MM40064742 |
42 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15918391
|
Emma Safety Footwear
MM40064743 |
43 |
132.00 EUR
1 pair |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
15928391
|
Emma Safety Footwear
MM40064744 |
44 |
131.00 EUR
1 pair |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
15938391
|
Emma Safety Footwear
MM40064745 |
45 |
109.18 EUR
1 pair |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
15948391
|
Emma Safety Footwear
MM40064746 |
46 |
109.18 EUR
1 pair |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
15958391
|
Emma Safety Footwear
MM40064747 |
47 |
109.18 EUR
1 pair |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
Description
- S1P-Safety shoe with sole plate for dry work
- Knitted Textile construction
- Polyurethane coated
- Hydro-Tec™ Antibacterial
- Metal-free
- Chrome-free lining
- ESD Certified
Specifications
EN ISO 20345:2011 | |
Black, Gray | |
Hydro-Tec™ Antibacterial | |
Composite |
Laces | |
Boston | |
Textile |