Learn More
Corning™ Bottle Top Dispensers (BTD) Cylinders
Tous les produits Corning Produits
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
2.5 mL
5 mL
10 mL
25 mL
50 mL
100 mL
missing translation for 'unitSize'
Each
Description
- Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
- Comes with four adapters with different diameters for universal use with multiple bottle sizes
- Telescopic suction tube can be adjusted smoothly to different bottle heights
- Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
- Every dispenser is individually tested and delivered with a certificate of performance
- For dispensing organic and inorganic liquids
Spécification
Spécification
| Volume (Metric) | 100 mL |
| For Use With (Application) | Dispensing Organic and Inorganic Liquids |
| Includes | Four adapters |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu