Learn More
Brady™ Thermal Transfer Conical Vial Labels
Product Code. 11831012
Click to view available options
Width (English):
1 in.
3.5 in.
Height (English):
1.5 in.
5.25 in.
Quantity:
2000/Pk.
3000/Pk.
Unit Size:
1 roll
Product Code. 11831012
Brand: Brady™ 062366
Only null left
Add to basket
Add to basket
Thermal Transfer Conical Vial Labels are made of self-laminating polyester (B-461), a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • Service temperature of -320 to 230° F (-196 to 110° C)
Specifications
Autoclavable Autoclavable
Certifications/Compliance RoHS, Pass 2002/95/EC
Description Thermal Transfer Printable Labels
Adhesive Type Acrylic, Permanent
Color White, Translucent
Display Consignment
Finish Matte
Format Label
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Thermal Transfer Printer Label
Sterility Non-sterile
For Use With (Application) Bottles, Conical tube, deep well plate, Laboratory identification, Petri dish, Tube, Vial
For Use With (Equipment) BBP81, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Labels per Row 1
Min. Operating Temperature (Metric) -196°C
Model THT-134-461-3
No. per Pack 3000 Labels/ Roll
Quantity 3000/Pk.
Shape Rectangle
Size 88.90 mm (W) x 38.10 mm (H)
Size (English) Panel 1.5 in H x 3.5 in W
Size (Metric) Panel 38.10 mm H x 88.90 mm W
Temperature Range (English) -321°F to +230°F
Temperature Range (Metric) -196°C to +110°C
Thickness (English) 0.0019 in.
Thickness (Metric) 0.048 mm
Vertical Repeat (Metric) 41.28 mm
Dimensions (LxW) 1.5 x 3.5 in.
Length (English) 1.5 in.
Width (English) 3.5 in.
Width (Metric) 88.9 mm
Width (Metric) Printable Area 38.10 mm
Height (English) 1.5 in.
Height (English) Printable Area 1.5 in.
Height (Metric) 38.10 mm
Height (Metric) Printable Area 31.75 mm
Alternate Part Number THT-134-461-3