Learn More
Brady™ BradyJet™ Continuous Polyester Labels
Brady™ BradyJet™ Continuous Polyester Labels are for use with BradyJet™ J2000 and are ideal for a wide range of identification applications.
Brand:  Brady™ 150031
 View more versions of this product
Product Code. 16110511
  • 328.00 EUR / Each
Estimated Shipment: 15-01-2025
Description
  • Constructed of indoor-use general purpose polyester
  • Features a specially-formulated topcoat for improved durability and color
  • Specially-formulated topcoat enhances colors and label appearance
  • Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
  • Topcoat protects print from smudging and harsh conditions
  • Inkjet-receptive labels for use with the BradyJet™ J2000 Color Label Printer
Specifications
Specifications
Acrylic
Matte
Polyester
BradyJet™ J2000 Printer
-40°C to +60°C
1.88 in.
White
Roll
Pipe and Valve Marking; Safety and Compliance ID; Visual Workplace Labeling; Warehouse Marking; Lockout Tagout Labeling; GHS and Chemical Labels; Arc Flash Labels
−40 to 140°F
100 ft. x 2.25 in.