Learn More
Brady™ BradyJet™ J5000 Repositionable Inkjet Vinyl Labels
The special aggressive adhesive on this Repositionable Inkjet Material stays in place when you need it to, but removes cleanly without leaving adhesive residue making it ideal for 5S and inventory applications.
Brand:  Brady™ 151112
 View more versions of this product
Product Code. 17429212
  • 496.00 EUR / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Description
  • Durable and flexible material making it ideal for areas where identification is frequently changed
  • Service temperature of -40 to 140°F (-40 to 60°C)
Specifications
Specifications
Acrylic
Matte
Repositionable
BradyJet™ J5000
-40°F to +140°F
White
Vinyl
Temporary Labeling
1
100 ft. x 6 in.