Learn More
VACUUBRAND™ VHCpro Suction Handset
Ergonomic light weight suction handset
Brand:  VACUUBRAND™ 20688061
167.23 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Product Code. 15293638
  • 187.00 EUR / Each
Estimated Shipment: 15-04-2025
Description
  • Autoclavable at 121°C and 2 barsv
  • Ready for use with 2.5 m flow through and flexible silicon tube, adapters for graduated, aspiration and pasteur pipettes, adapter for pipette tips, wall holder for standby-position and clips with adhesive surface to guide the suction tube into the biosafety cabinet neatly
  • Permanent suction mode for continuous flow
  • Mechanism of the VHCpro handset is not in contact with contaminated fluids and enables reliable, leak free working
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VHCpro Suction Handset
Product Title
VACUUBRAND™ VHCpro Suction Handset > VHCpro Suction Handset