Learn More
MP Biomedicals Briggs Chick Salt Mixture A
Salt mixture high in calcium phosphate, calcium chloride.
Brand: MP Biomedicals 0290133250
This item is not returnable.
View return policy
Description
- Animal diet high in salts
- Salt mixture high in calcium phosphate and calcium chloride
Specifications
Not Autoclavable | |
Single-use | |
Non-sterile | |
1 ft. |
Animal Diet | |
50 kg | |
Room Temperature | |
0.3048 m |
Product Content Correction
Product Title
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction