missing translation for 'onlineSavingsMsg'
Learn More

BRL 37344, sodium salt, Tocris Bioscience™

Product Code. 18064582
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

β3 agonist

Potent and selective β3 adrenoceptor agonist (Ki values are 287, 1750 and 1120 nM for β3, β1 and β2 receptors respectively).

Chemical Identifiers

CAS 127299-93-8
Molecular Formula C19H21ClNNaO4
Molecular Weight (g/mol) 385.82
InChI Key SNJIJYKMYQRHRC-QRGZVCNKSA-M
Synonym 4-s-2-s-2-3-chlorophenyl-2-hydroxyethyl amino propyl phenoxy acetic acid sodium salt, sodium 2-4-2s-2-2s-2-3-chlorophenyl-2-hydroxyethyl amino propyl phenoxy acetate
PubChem CID 91667707
IUPAC Name sodium;2-[4-[(2R)-2-[[(2S)-2-(3-chlorophenyl)-2-hydroxyethyl]amino]propyl]phenoxy]acetate
SMILES CC(CC1=CC=C(C=C1)OCC(=O)[O-])NCC(C2=CC(=CC=C2)Cl)O.[Na+]

Specifications

Quantity 10 mg
Formula Weight 385.82
Percent Purity >98%
Chemical Name or Material BRL 37344, sodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.