missing translation for 'onlineSavingsMsg'
Learn More

Brombuterol-(tert-butyl-d9) hydrochloride, Honeywell Fluka™

Product Code. 15967412
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY

Specifications

CAS 1353867-94-3
Packaging Glass Bottle
Quantity 10 mg
Molecular Formula C12D9H9Br2N2O · HCl
MDL Number MFCD20264918
Molecular Weight (g/mol) 411.61
Grade Analytical Standard
For Use With (Application) Beta-agonists and Antagonists
Physical Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.