missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromide-Bromate, 0.05mol/l Solution, Scharlab
Used in analytical chemistry as a titrant in volumetric analysis. Scharlab Bromide-Bromate, 0.05mol/l Solution is prepared according to ASTM D5776-99.
Brand: Scharlab BR00701000
This item is not returnable.
View return policy
Description
This volumetric solution was checked by means of potentiometric methods using a sodium thiosulfate standard solution, that was also checked against Scharlau's potassium iodate volumetric standard. Scharlau's volumetric standards are directly traceable to the Standard Reference Materials from NIST (National Institute of Standards and Technology, USA).
- Molecular Formula: Br- / BrO3-
- Factor: 0.999–1.001
Specifications
Solution | |
Br- / BrO3- | |
∼1.018g/cm3 |
1 L | |
ASTM |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Bromide-bromate, solution 0,05 mol/ (0,1 N), according to ASTM D5776-99
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1B
Hazard Statement
- H350-May cause cancer.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Potassium bromate