missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Green, Pure, Indicator Grade, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Specifications
Specifications
| Melting Point | 225°C |
| CAS | 76-60-8 |
| Molecular Formula | C21H14Br4O5S |
| MDL Number | 5874 |
| Formula Weight | 698.01g/mol |
| Packaging | Glass Jar |
| Color | Beige |
| Quantity | 5 g |
| Physical Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction