Learn More
Bromocyclohexane, 98%, Thermo Scientific Chemicals
27.10 EUR - 243.00 EUR
Chemical Identifiers
CAS | 108-85-0 |
---|---|
Molecular Formula | C6H11Br |
Molecular Weight (g/mol) | 163.06 |
MDL Number | MFCD00003819 |
InChI Key | AQNQQHJNRPDOQV-UHFFFAOYSA-N |
Synonym | cyclohexyl bromide, cyclohexane, bromo, 1-bromocyclohexane, unii-ya0ums4rny, bromo-cyclohexane, ya0ums4rny, cyclohexylbromide, bromocylcohexane, bromocyclohexane, ksc178g7n |
PubChem CID | 7960 |
IUPAC Name | bromocyclohexane |
SMILES | BrC1CCCCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494663
|
Thermo Scientific Alfa Aesar
A11593.22 |
100 g |
27.10 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404673
|
Thermo Scientific Alfa Aesar
A11593.36 |
500 g |
71.30 EUR
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414673
|
Thermo Scientific Alfa Aesar
A11593.0E |
2500 g |
243.00 EUR
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBromocyclohexane has been used to study the ionization of equatorial and axial conformational isomers of the chair-bromocyclohexane using vacuum ultraviolet mass-analyzed threshold ionization spectroscopy.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Air & Light Sensitive. Store away from strong bases.
Chemical Identifiers
108-85-0 | |
163.06 | |
AQNQQHJNRPDOQV-UHFFFAOYSA-N | |
7960 | |
BrC1CCCCC1 |
C6H11Br | |
MFCD00003819 | |
cyclohexyl bromide, cyclohexane, bromo, 1-bromocyclohexane, unii-ya0ums4rny, bromo-cyclohexane, ya0ums4rny, cyclohexylbromide, bromocylcohexane, bromocyclohexane, ksc178g7n | |
bromocyclohexane |
Specifications
108-85-0 | |
1.335 | |
62°C (143°F) | |
1.495 | |
1098373 | |
14,2730 | |
Soluble in alcohol. Insoluble in water. | |
BrC1CCCCC1 | |
163.06 | |
163.06 | |
Bromocyclohexane |
-57°C | |
166°C to 167°C | |
C6H11Br | |
MFCD00003819 | |
Light sensitive | |
cyclohexyl bromide, cyclohexane, bromo, 1-bromocyclohexane, unii-ya0ums4rny, bromo-cyclohexane, ya0ums4rny, cyclohexylbromide, bromocylcohexane, bromocyclohexane, ksc178g7n | |
AQNQQHJNRPDOQV-UHFFFAOYSA-N | |
bromocyclohexane | |
7960 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
missing translation for 'einecsNumber' : 203-622-2
missing translation for 'rtecsNumber' : GU7171700
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only