Learn More
Emma Safety Footwear Bryce Safety Shoes
The impenetrable sole protects your feet from sharp and metal objects while the steel toe cap and reinforced PU toe cover offer protection from falling objects. Microfiber construction remains flexible to stand up to cement and resists corrosive substances.
Specifications
Certifications/Compliance | EN ISO 20345:2011 |
---|---|
Closure Type | Laces |
Color | Black |
Design | Bryce |
Liner Material | Hydro-Tec™ Sanitized Silver |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15918351
|
Emma Safety Footwear
MM33084837 |
37 |
N/A
|
N/A | |||||
15928351
|
Emma Safety Footwear
MM33084838 |
38 |
N/A
|
N/A | |||||
15938351
|
Emma Safety Footwear
MM33084839 |
39 |
N/A
|
N/A | |||||
15948351
|
Emma Safety Footwear
MM33084840 |
40 |
N/A
|
N/A | |||||
15958351
|
Emma Safety Footwear
MM33084841 |
41 |
N/A
|
N/A | |||||
15968351
|
Emma Safety Footwear
MM33084842 |
42 |
N/A
|
N/A | |||||
15978351
|
Emma Safety Footwear
MM33084843 |
43 |
N/A
|
N/A | |||||
15988351
|
Emma Safety Footwear
MM33084844 |
44 |
N/A
|
N/A | |||||
15998351
|
Emma Safety Footwear
MM33084845 |
45 |
N/A
|
N/A | |||||
15908361
|
Emma Safety Footwear
MM33084846 |
46 |
N/A
|
N/A | |||||
15918361
|
Emma Safety Footwear
MM33084847 |
47 |
N/A
|
N/A | |||||
15928361
|
Emma Safety Footwear
MM33084848 |
48 |
N/A
|
N/A | |||||
15938361
|
Emma Safety Footwear
MM33084849 |
49 |
N/A
|
N/A | |||||
Description
- S3-Safety shoe with sole plate for wet work
- GripForce™ Duo Heat Resistant Outsole
- Microfiber construction
- 3M™ Reflective Striping
- Hydro-Tec™ Sanitized Silver
Specifications
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
Steel |
Laces | |
Bryce | |
Microfiber |