Learn More
Paragon Scientific™ BS/IP/SL(S) Viscometer for Transparent Liquids
Product Code. 16000400
Click to view available options
Viscosity Range:
1.05 min
2.1 to 3 cSt
20 to 100 cSt
200 to 1000 cSt
2000 to 10,000 cSt
3.8 to 10 cSt
6 to 30 cSt
60 to 300 cSt
600 to 3000 cSt
Unit Size:
Each
Product Code. 16000400
Brand: Paragon Scientific™ SHORT07NC
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Viscosity Range 200 to 1000 cSt
Calibrated No
Approximate Constant 1
ASTM Numbers D 445, D 446
Certifications/Compliance ASTM D 445, D 446 and ISO 3104, 3105
Size 7
4 week lead-time