Learn More
Eurofins DiscoverX BTK Protein, Active
Description
BTK (Bruton's Tyrosine Kinase) is a member of the Tec family kinases which are important components of both antigen receptor signaling and other cell surface receptors, resulting in regulation of the actin cytoskeleton, adhesion, migration, and transcriptional activation. BTK is found in all cells of the hematopoietic lineage except plasma and T cells, and is required for normal B cell development and signal transduction through cell surface molecules. Mutations in the btk gene result in aberrant B cell development, leading to the X-linked agammaglobulinemia phenotype in humans and to the less severe X-linked immunodeficiency in mice. BTK inhibitors may be of benefit in the treatment of leukemias and lymphomas.
GenBank NM_000061
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 78.4 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥53% |
| Protein | BTK, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.