Learn More
Thermo Scientific™ Buckets and Lids for Thermo Scientific Sorvall™ Legend™ Mach 1.6™ 4-Place Swinging Bucket Rotor 75-002-000
Shop All Thermo Scientific Products
Click to view available options
Type:
Bucket
Cap
Capacity:
150 mL
400 mL
Not Applicable
Unit Size:
Each
Description
For use with Sorvall Legend Mach 1.6/1.6R Centrifuges when equipped with 4-Place Swinging Bucket Rotor 75-002-000.
Choose rectangular or round buckets.
- Rectangular Bucket (75-002-001) capacity: 150mL
- Round Bucket (75-002-002) capacity: 400mL
- Aerosol-tight lids are suitable for biocontainment use
Order Info
Buckets and lids are sold separately. Buckets are sold individually; order correct quantity for your rotor. Lids are sold in pairs.
Specifications
Specifications
| For Use With (Equipment) | TTH-400 swing bucket rotor |
| Capacity | 150 mL |
| Type | Bucket |
| Description | Rectangular Bucket |