missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 1.00, Chem-Lab
Designed for pH measurement and maintenance in highly acidic samples. Buffer Solution, pH 1.00, Chem-Lab is a mixture of potassium chloride and hydrochloric acid with a pH level of 1.00 at 20°C. Solution is traceable to standard reference materials (SRM) from NIST.
Brand: Chem Lab CL03.0203.1000
This item is not returnable.
View return policy
Description
pH Level ± 0.02 at specified temperature:
- pH 0.94 at 0°C
- pH 0.99 at 10°C
- pH 1.00 at 20°C
- pH 1.00 at 25°C
- pH 1.00 at 30°C
- pH 1.01 at 40°C
- pH 1.01 at 50°C
- pH 1.01 at 60°C
- pH 1.01 at 70°C
- pH 1.02 at 80°C
- pH 1.02 at 90°C
Specifications
7447-40-7, 7647-01-0, 7732-18-5 | |
Colorless | |
1.00 (at 20°C to 30°C) | |
1 L | |
KCl-HCl | |
Yes |
0°C | |
1.01g/mL | |
Liquid | |
pH Buffer | |
Odorless | |
Water Soluble |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction