Learn More
Buffer Solution pH 6.5, Fisher Chemical™
Product Code. 12910704
Click to view available options
Quantity:
1000 mL
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
pH 6.5
Physical Form Liquid
Quantity 1000 mL
Solution Type Buffer