missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 7, Scharlab

Product Code. p-7083572
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 11329903

Brand: Scharlab SO10070250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to fit the pH of the reaction media, for calibrating pH meters and in analytical chemistry.

Standard buffer solutions are prepared using gravimetric and volumetric procedures. The batch value is determined by measurement with a combination glass electrode against five-point calibration according to DIN 19268. This pH buffer solution is traceable to Standard Reference Material from NIST.

Specifications

Melting Point -5°C
Boiling Point 109°C
Density 1.01g/cm3
pH 7.00
Physical Form Solution
Quantity 250 mL
Formulation Per liter: 3.54g Potassium dihydrogen phosphate, 14.7g Disodium hydrogen phosphate. Contains preservative.
Traceability to NIST Traceable to NIST
Solubility Information Miscible in water (20°C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.