missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution, Potassium Dihydrogen Phosphate/di-Sodium Hydrogene Phosphate, pH 7.00, Fisher Chemical™
BIOPHARMA

Product Code. 12990644 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
This item is not returnable. View return policy

Product Code. 12990644

Brand: Fisher Chemical SOL 005 1L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Chemical Name or Material Buffer solution, Potassium Dihydrogen Phosphate/di-Sodium Hydrogen Phosphate
pH 7
Quantity 1000 mL
Packaging Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.