Learn More
Uvex Busingess Casual 95388 Insole - Width 11
Shop All Uvex Products
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Description
- Full surface foot shock absorption
- Highly breathable and moisture-absorbent material
- Perforation for improved breathability
- Shock absorption zones support natural foot movement
- Skin compatibility and pleasant surface structure
- Supports foot arch
- Article No. 9538.8
- Size: 38 to 48
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | Uvex Business Casual |
| Material | Microvelour |
| Size | 41 |
| Disposable | Reusable |
| Gender | Unisex |
| Fit | Standard width |
| Latex Free | Yes |
| Waterproof | No |