missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint Gobain Life Sciences™ C-Flex™ 072 TPE Tubing
Biocompatible, sealable, and weldable TPE tubing.
419.00 EUR - 1995.00 EUR
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Thermoplastic |
For Use With | Biopharmaceutical Applications |
Color | Opaque |
Product Type | Tubing |
Product Code | Brand | Diameter (Metric) Outer | Maximum Pressure | Overall Length (Metric) | Thickness (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Maximum Pressure | Overall Length (Metric) | Thickness (Metric) | Price | Quantity & Availability | ||
17993711
|
Saint Gobain Life Sciences™
072-125-2 |
6.4 mm | 32 psi (at 19°C) | 15 m | 1.6 mm |
460.00 EUR
120m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17913501
|
Saint Gobain Life Sciences™
072-250-2M |
9.5 mm | 20 psi (at 19°C) | 107 m | 1.6 mm |
428.00 EUR
107m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17923421
|
Saint Gobain Life Sciences™
072-250-2S |
9.5 mm | 20 psi (at 19°C) | 305 m | 1.6 mm |
1995.00 EUR
305m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17963581
|
Saint Gobain Life Sciences™
072-188-3 |
9.5 mm | 24 psi (at 19°C) | 15 m | 2.4 mm |
419.00 EUR
75m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17993491
|
Saint Gobain Life Sciences™
072-188-3M |
9.5 mm | 42 psi (at 19°C) | 107 m | 2.4 mm |
580.00 EUR
107m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17903701
|
Saint Gobain Life Sciences™
072-250-3 |
11.2 mm | 26 psi (at 19°C) | 15 m | 2.4 mm |
621.00 EUR
90m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17933421
|
Saint Gobain Life Sciences™
072-313-3M |
12.7 mm | 21 psi (at 19°C) | 61 m | 2.4 mm |
478.00 EUR
61m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17913561
|
Saint Gobain Life Sciences™
072-313-3S |
12.7 mm | 21 psi (at 19°C) | 152 m | 2.4 mm |
1130.00 EUR
152m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17953421
|
Saint Gobain Life Sciences™
072-375-3 |
14.3 mm | 19 psi (at 19°C) | 15 m | 2.4 mm |
544.00 EUR
60m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17993501
|
Saint Gobain Life Sciences™
072-375-4 |
15.9 mm | 24 psi (at 19°C) | 15 m | 3.2 mm |
800.00 EUR
60m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17943511
|
Saint Gobain Life Sciences™
072-375-4M |
15.9 mm | 24 psi (at 19°C) | 46 m | 3.2 mm |
528.00 EUR
46m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17913511
|
Saint Gobain Life Sciences™
072-375-4S |
15.9 mm | 24 psi (at 19°C) | 107 m | 3.2 mm |
1230.00 EUR
107m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17973511
|
Saint Gobain Life Sciences™
072-500-4S |
19.05 mm | 17 psi (at 19°C) | 76 m | 3.2 mm |
1120.00 EUR
76m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17973581
|
Saint Gobain Life Sciences™
072-500-3 |
19.1 mm | 14 psi (at 19°C) | 15 m | 2.4 mm |
518.00 EUR
45m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
C-Flex™ biopharmaceutical tubing is the original patented thermoplastic elastomer tubing specifically designed to meet the critical demands of pharmaceutical and biopharmaceutical applications for fluid processing.
- Specifically designed for aseptic welding connections and sealing disconnection
- Suitable for sterilization by gamma and x-ray irradiation, as well as autoclave
- Moldable, bondable, and formable, making it ideal for single-use assemblies, overmolds, tubing, and bag manifolds
- Manufactured from pharmaceutical-grade thermoplastic elastomer
- Sealable and weldable either pre- or post- sterilization
- Complies with USP Class VI and ISO standards
- Remains flexible from -45°C to +135°C (-50°F to +275°F)
- Significantly less permeable than silicone
- Animal-Derived Component-Free (ADCF)
- Very high durability in peristaltic pump applications
- Typical applications include: Aseptic Sealing Disconnections; Aseptic Welding Connections; Fluid Transfer in Pharmaceutical, Biopharmaceutical, and/or Laboratory Settings; Single-use Assemblies
Specifications
Autoclavable | |
Biopharmaceutical Applications | |
Tubing |
Thermoplastic | |
Opaque |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title