Learn More
Uvex™ C300 Dry Cut Protection Gloves
Click to view available options
Size:
8
9
10
11
Description
- Gloves provide level 3/C blade cut resistance, and very high tear resistance
- High-performance vinyl (HPV) dots on the palm enhance grip capabilities in dry environments
- Uvex Climazone technology minimizes excessive warmth and moisture
- Highly flexible with very good tactile sensation
- Silicone-free according to the imprint test
- 3D Ergo technology ensures a secure fit
Specifications
Specifications
Coating Material | HPV Dots |
Color | Gray, Green |
Material | Bamboo Viscose, HPPE, Polyamide, Glass |
Size | 8 |
EN388 Standard | XX4XC |
Glove Cuff Style | Knit |
Series | C300 Dry |