Learn More
WTW™ Cadmium Test Kits
Obtain precision measurements for waste water, seepage water, and process water with CombiCheck Cadmium (Cd) cell test kits.
Brand: WTW™ 250314
This item is not returnable.
View return policy
Description
- For use with PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
- For waste water, seepage water, process water
- CombiCheck Cadmium (Cd) cell test kit product type
- Cadion derivative method
- Photometric test results
Specifications
Cadion derivative | |
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers | |
25 | |
Photometric |
For waste water, seepage water, process water | |
Certificates | |
25 Tests | |
CombiCheck Cadmium (Cd) cell test kit |
Safety and Handling
Product Identifier
Hazard Category
Supplemental information
- Cadmium Cell Test Method: photometric 0.025 - 1.000 mg/l Cd Spectroquant
Hazard Category
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Supplemental information
- MIXTURE LIST-Contains : Triton-X-100, disodium tetraborate; N,N-dimethylformamide