missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Carbonate, ≥99.0%, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Melting Point | 825°C |
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Linear Formula | CaCO3 |
UN Number | NONH for all modes of transport |
Beilstein | 8008338 |
IUPAC Name | calcium carbonate |
Formula Weight | 100.09g/mol |
Percent Purity | ≥99.0% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'