missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, Dihydrate, MP Biomedicals™

Product Code. 11465092 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Calcium chloride, Dihydrate Cell Culture Reagent, ∽99%, White Powder

  • Calcium chloride is a commonly used reagent in biochemistry.
  • It plays important roles in many biological processes, including signal transduction, muscle contraction, and maintenance of cell membrane and cell wall stability.
  • Calcium chloride is used in the preparation and transformation of competent E. coli and in the transfection of eukaryotic cells with either plasmid DNA or high molecular weight genomic DNA. has been used in the stabilization and two dimensional crystallization of the NADH-ubiquinone oxidoreductase (complex I) from Escherichia coli.
  • It is also used as a dessicant, coagulant in rubber manufacturing and as an accelerator in the concrete industry.
  • Calcium Chloride is a salt of calcium and chloride.
  • It behaves as a typical ionic halide, and is solid at room temperature
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00149613
InChI Key LLSDKQJKOVVTOJ-UHFFFAOYSA-L
Synonym Calcium dichloride dihydrate

Specifications

Melting Point 176°C (decomposition)(literature)
pH 4 to 9.5 (5% aq. soln.)
Physical Form Powder
Quantity 500 g
Molecular Formula CaCl2H4O2
Solubility Information Soluble in water - 294mg/mL (clear, colorless solution) with heat liberation, ethanol, acetic acid, acetone.
Molecular Weight (g/mol) 147.01
Grade ∽99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.