Learn More
WTW™ Calcium Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand:  WTW™ 250428
 View more versions of this product
Product Code. 10134211
  • 237.00 EUR / 10mL
Explore more special offers
Specifications
Eosin/phenanthroline
For PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
100
100 Tests
Silver (Ag) reagent test kit
For waste water, drinking water
Certificates
5 to 160 mg/L
Photometric
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Calcium Test Method: photometric 5 - 160 mg/l Ca
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : diethylene glycol