missing translation for 'onlineSavingsMsg'
Learn More

Calibration Buffer pH: 9.18, NIST Traceable, Sachets, Hanna Instruments™

Product Code. 11637020
Click to view available options
Quantity:
20 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11637020

Brand: Hanna Instruments HI70009C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color Colorless
pH 9.18
Physical Form Liquid
Quantity 20 mL
Solution Type Buffer
Formulation Aqueous Buffer Solution
Odor Odorless
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.