missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calyculin A, >98%, MP Biomedicals™
Click to view available options
Quantity:
10 μg
50 μg
Spezifikation
Spezifikation
Assay Percent Range | >98% |
Quantity | 10 μg |
Formula Weight | 1009.18 |
Physical Form | Lyophilized Powder |
Chemical Name or Material | Calyculin A |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'