Learn More
Fisherbrand™ Huygens Eyepiece
Use this eyepiece with compound microscopes. Fisherbrand™ Huygens Eyepieces are ergonomically designed for everyday use.
Brand:  Fisherbrand™ 9554.1019
Product Code. 12378518
  • 464.00 EUR / Each
Estimated Shipment: 27-03-2025
Description
For compound microscopes FB69019 and FB69020
  • Affordable compound microscopes for use by students and professionals
  • Includes powerful low voltage illumination with intensity controller, a built in mechanical stage and four
  • achromatic objectives 4x, 10x, 40x and 100x oil (100x for FB69020 only)
  • 10x WF eyepieces
  • Pre-centered Abbe condenser
  • Includes immersion oil 10mL (FB69020 only), blue filter, dust cover and instruction manual
Specifications
Specifications
Huygens Eyepiece
Compound microscopes FB69019 and FB69020